siRNA Details
Gene Symbol: EPHA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005233.5 | NP_005224.2 | 11, 12 | 2292 | ||
XM_005264715.2 | XP_005264772.1 | 11, 12 | 2214 | |||
XM_005264716.2 | XP_005264773.1 | 11, 12 | 2287 | |||
GenBank mRNA | AF213459.1 | - | 11, 12 | 2292 | ||
AK291411.1 | - | 11, 12 | 2125 | |||
BC063282.1 | - | 11, 12 | 2196 | 30347891 | ||
M83941.1 | - | 11, 12 | 2167 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
