siRNA Details
Gene Symbol: SIK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281748.1 | NP_001268677.1 | 21 | 3267 | ||
NM_001281749.1 | NP_001268678.1 | 20 | 3143 | |||
NM_025164.4 | NP_079440.3 | 20 | 3323 | |||
XM_005271481.2 | XP_005271538.1 | 21 | 3505 | |||
XM_005271482.4 | XP_005271539.2 | 20 | 3361 | |||
XM_005271484.3 | XP_005271541.2 | 20 | 3181 | |||
XM_005271485.3 | XP_005271542.1 | 8 | 1843 | |||
XM_011542721.2 | XP_011541023.1 | 21 | 3467 | |||
XM_011542722.1 | XP_011541024.1 | 21 | 3325 | |||
XM_011542723.2 | XP_011541025.1 | 21 | 3348 | |||
XM_011542724.2 | XP_011541026.1 | 20 | 3202 | |||
XM_011542725.2 | XP_011541027.1 | 19 | 3085 | |||
XM_017017424.1 | XP_016872913.1 | 19 | 3037 | |||
XM_017017425.1 | XP_016872914.1 | 21 | 3543 | |||
XM_017017426.1 | XP_016872915.1 | 19 | 3417 | |||
XM_017017427.1 | XP_016872916.1 | 16 | 2820 | |||
GenBank mRNA | AB209327.1 | - | 13 | 2333 | ||
AK022302.1 | - | 2 | 878 | |||
AK122838.1 | - | 6 | 2585 | |||
AY598338.1 | - | 2 | 875 | |||
BC035583.2 | - | 3 | 1153 | |||
BC063887.1 | - | 6 | 1336 | |||
BC086858.1 | - | 2 | 878 | |||
BC128510.1 | - | 20 | 3005 | 40081639 | ||
BC128511.1 | - | 21 | 3123 | 40081640 |
