siRNA Details
Gene Symbol: ZAP70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079.3 | NP_001070.2 | 4 | 659 | ||
XM_017004867.1 | XP_016860356.1 | 6 | 2060 | |||
XM_017004868.1 | XP_016860357.1 | 6 | 2060 | |||
XM_017004869.1 | XP_016860358.1 | 6 | 2060 | |||
XM_017004870.1 | XP_016860359.1 | 6 | 2060 | |||
GenBank mRNA | AK303341.1 | - | 4 | 651 | ||
AK310628.1 | - | 4 | 645 | |||
AK310637.1 | - | 4 | 651 | |||
BC039039.1 | - | 3 | 313 | 5168073 | ||
BC053878.1 | - | 4 | 635 | 5757161 | ||
L05148.1 | - | 4 | 661 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
