siRNA Details
Gene Symbol: SRPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278273.1 | NP_001265202.1 | 11 | 1022 | ||
NM_182691.2 | NP_872633.1 | 9 | 1075 | |||
NM_182692.2 | NP_872634.1 | 10 | 998 | |||
XM_005250549.3 | XP_005250606.2 | 11 | 1257 | |||
XM_005250550.3 | XP_005250607.1 | 10 | 1002 | |||
XM_005250551.4 | XP_005250608.1 | 11 | 1050 | |||
XM_006716098.2 | XP_006716161.1 | 11 | 1017 | |||
XM_011516536.2 | XP_011514838.1 | 9 | 1013 | |||
XM_011516538.2 | XP_011514840.1 | 10 | 886 | |||
XM_017012560.1 | XP_016868049.1 | 11 | 1072 | |||
XM_017012561.1 | XP_016868050.1 | 10 | 922 | |||
XM_017012562.1 | XP_016868051.1 | 11 | 1256 | |||
XM_017012563.1 | XP_016868052.1 | 11 | 1257 | |||
XM_017012564.1 | XP_016868053.1 | 9 | 1061 | |||
XM_017012565.1 | XP_016868054.1 | 11 | 1021 | |||
XM_017012566.1 | XP_016868055.1 | 11 | 1058 | |||
GenBank mRNA | AK093420.1 | - | 1 | 797 | ||
AY354201.1 | - | 10 | 1063 | |||
BC035214.1 | - | 9 | 994 | 4778251 | ||
BC068547.1 | - | 11 | 1022 | 5265599 | ||
U88666.1 | - | 9 | 1025 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
