siRNA Details
Gene Symbol: TAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138923.3 | NP_620278.1 | 4, 5 | 600 | ||
XM_005262297.4 | XP_005262354.1 | 4, 5 | 619 | |||
GenBank mRNA | D90359.1 | - | 4, 5 | 575 | ||
X07024.1 | - | 3, 4 | 687 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
