siRNA Details
Gene Symbol: EGFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005228.3 | NP_005219.2 | 12 | 1647 | ||
NM_201282.1 | NP_958439.1 | 12 | 1647 | |||
NM_201284.1 | NP_958441.1 | 12 | 1647 | |||
GenBank mRNA | AB209442.1 | - | 12 | 1546 | ||
AF125253.1 | - | 12 | 1646 | |||
AK225422.1 | - | 10 | 1298 | |||
AK290352.1 | - | 12 | 1447 | |||
AK294750.1 | - | 12 | 1425 | |||
AY550966.1 | - | 12 | 1447 | |||
AY698023.1 | - | 12 | 1401 | |||
BC094761.1 | - | 11 | 1523 | 30528231 | ||
BC118665.1 | - | 7 | 783 | |||
BC128419.1 | - | 6 | 586 | |||
DQ088980.1 | - | 12 | 1447 | |||
GU255993.1 | - | 12 | 1444 | |||
HQ912715.1 | - | 11 | 1512 | |||
K03193.1 | - | 12 | 1560 | |||
X00588.1 | - | 12 | 1587 | |||
X00663.1 | - | 9 | 975 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
