siRNA Details
Gene Symbol: CAMK1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020439.2 | NP_065172.1 | 4 | 362 | ||
XM_017001866.1 | XP_016857355.1 | 4 | 505 | |||
XM_017001867.1 | XP_016857356.1 | 4 | 345 | |||
GenBank mRNA | AF428261.1 | - | 4 | 332 | ||
AK095713.1 | - | 4 | 505 | |||
AK289675.1 | - | 4 | 345 | |||
AL049688.1 | - | 3 | 277 | |||
AY212935.1 | - | 3 | 268 | |||
BC032787.1 | - | 4 | 329 | 5179957 | ||
BG715920.1 | - | 4 | 366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
