siRNA Details
Gene Symbol: FGFR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000142.4 | NP_000133.1 | 8 | 1224 | ||
XM_006713872.1 | XP_006713935.1 | 8 | 1237 | |||
XM_006713873.1 | XP_006713936.1 | 8 | 1237 | |||
XM_011513420.1 | XP_011511722.1 | 8 | 1237 | |||
XM_011513422.1 | XP_011511724.1 | 8 | 1237 | |||
GenBank mRNA | AB209441.1 | - | 8 | 1229 | ||
AF369211.1 | - | 4 | 389 | |||
AF369212.1 | - | 4 | 365 | |||
AF369213.1 | - | 4 | 326 | |||
HM244683.1 | - | 8 | 1119 | |||
KJ534834.1 | - | 7 | 968 | |||
M58051.1 | - | 7 | 1007 | |||
M64347.1 | - | 6 | 743 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
