siRNA Details
Gene Symbol: CDK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320918.1 | NP_001307847.1 | 5 | 580 | ||
NM_001786.4 | NP_001777.1 | 5 | 568 | |||
XM_005270303.3 | XP_005270360.1 | 5 | 650 | |||
GenBank mRNA | AK291939.1 | - | 5 | 505 | ||
BC014563.1 | - | 4 | 439 | 2967728 | ||
BC107750.1 | - | 5 | 529 | 6710672 | ||
BF685285.1 | - | 5 | 628 | |||
BG723076.1 | - | 5 | 572 | |||
BT007004.1 | - | 4 | 426 | |||
CR933728.1 | - | 4 | 2795 | |||
X05360.1 | - | 5 | 552 | |||
Y00272.1 | - | 5 | 552 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
