siRNA Details
Gene Symbol: CASP1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131905.1 | 2, 3 | 355 | |||
GenBank mRNA | AB231723.1 | - | 1, 2 | 95 | ||
CB985891.1 | - | 2, 3 | 358 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
