siRNA Details
Gene Symbol: LTB4R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164692.2 | NP_001158164.1 | 2 | 899 | ||
NM_019839.4 | NP_062813.2 | 2 | 843 | |||
GenBank mRNA | AB029892.1 | - | 1 | 728 | ||
AB032558.1 | - | 2 | 809 | |||
AB044402.1 | - | 1 | 728 | |||
AF277230.1 | - | 1 | 2439 | |||
AK304169.1 | - | 2 | 835 | |||
BC038437.1 | - | 2 | 879 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
