siRNA Details
Gene Symbol: HGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000601.5 | NP_000592.3 | 6 | 896 | ||
NM_001010931.2 | NP_001010931.1 | 6 | 896 | |||
NM_001010932.2 | NP_001010932.1 | 6 | 881 | |||
NM_001010933.2 | NP_001010933.1 | 6 | 881 | |||
XM_006715956.2 | XP_006716019.1 | 7 | 859 | |||
XM_011516115.2 | XP_011514417.1 | 7 | 844 | |||
XM_017012097.1 | XP_016867586.1 | 7 | 859 | |||
XM_017012098.1 | XP_016867587.1 | 7 | 844 | |||
GenBank mRNA | AK291674.1 | - | 6 | 831 | ||
AK313694.1 | - | 6 | 731 | |||
BC022308.1 | - | 6 | 730 | |||
BC130284.1 | - | 6 | 744 | 40146315 | ||
BC130286.1 | - | 6 | 744 | 40146317 | ||
L02931.1 | - | 6 | 729 | |||
M29145.1 | - | 6 | 770 | |||
M55379.1 | - | 6 | 654 | |||
M60718.1 | - | 6 | 741 | |||
M73239.1 | - | 6 | 734 | |||
M73240.1 | - | 6 | 719 | |||
M77227.1 | - | 6 | 751 | |||
X16323.1 | - | 6 | 803 | |||
X57574.1 | - | 6 | 746 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
