siRNA Details
Gene Symbol: HGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000601.5 | NP_000592.3 | 12 | 1646 | ||
NM_001010932.2 | NP_001010932.1 | 12 | 1631 | |||
XM_006715956.2 | XP_006716019.1 | 13 | 1609 | |||
XM_011516115.2 | XP_011514417.1 | 13 | 1594 | |||
GenBank mRNA | AK291674.1 | - | 12 | 1581 | ||
AK313694.1 | - | 12 | 1481 | |||
BC130284.1 | - | 12 | 1494 | 40146315 | ||
BC130286.1 | - | 12 | 1494 | 40146317 | ||
M29145.1 | - | 12 | 1520 | |||
M55379.1 | - | 12 | 1404 | |||
M60718.1 | - | 12 | 1491 | |||
M73239.1 | - | 12 | 1484 | |||
M73240.1 | - | 12 | 1469 | |||
X16323.1 | - | 12 | 1553 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
