siRNA Details
Gene Symbol: CAMK1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020439.2 | NP_065172.1 | 11, 12 | 1436 | ||
XM_017001866.1 | XP_016857355.1 | 11, 12 | 1579 | |||
XM_017001867.1 | XP_016857356.1 | 11, 12 | 1430 | |||
GenBank mRNA | AF428261.1 | - | 11, 12 | 1406 | ||
AK095713.1 | - | 11, 12 | 1579 | |||
AL049688.1 | - | 10, 11 | 1351 | |||
BC032787.1 | - | 11, 12 | 1403 | 5179957 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
