siRNA Details
Gene Symbol: MBTPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003791.3 | NP_003782.1 | 13 | 2281 | ||
XM_017023816.1 | XP_016879305.1 | 12 | 2030 | |||
XM_017023817.1 | XP_016879306.1 | 14 | 2284 | |||
GenBank mRNA | AK091212.1 | - | 2 | 992 | ||
AK291773.1 | - | 13 | 2164 | |||
AK299067.1 | - | 12 | 1506 | |||
BC114555.1 | - | 13 | 2167 | 40054075 | ||
BC114961.1 | - | 13 | 2167 | 40054074 | ||
D42053.1 | - | 13 | 2244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
