siRNA Details
Gene Symbol: DPP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK289967.1 | - | 9, 10 | 980 | ||
AY172661.3 | - | 9, 10 | 860 | |||
BC030832.1 | - | 9, 10 | 1317 | 4811771 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
