siRNA Details
Gene Symbol: MASP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001879.5 | NP_001870.3 | 9 | 1546 | ||
NM_139125.3 | NP_624302.1 | 9 | 1546 | |||
NR_033519.1 | 8 | 1314 | ||||
XM_011512989.2 | XP_011511291.1 | 9 | 1540 | |||
XM_011512990.2 | XP_011511292.1 | 9 | 1540 | |||
XM_017006869.1 | XP_016862358.1 | 10 | 1624 | |||
XM_017006870.1 | XP_016862359.1 | 8 | 1512 | |||
XM_017006871.1 | XP_016862360.1 | 9 | 1540 | |||
XM_017006872.1 | XP_016862361.1 | 9 | 1399 | |||
GenBank mRNA | AF284421.1 | - | 9 | 1246 | ||
AK291157.1 | - | 9 | 1382 | |||
AK291686.1 | - | 9 | 1224 | |||
AK304334.1 | - | 8 | 1024 | |||
AL708781.1 | - | 1 | 30 | |||
BC106945.1 | - | 9 | 1206 | 40034551 | ||
BC106946.1 | - | 9 | 1206 | 40034552 | ||
BX641029.1 | - | 1 | 1421 | |||
BX647237.1 | - | 8 | 1029 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
