siRNA Details
Gene Symbol: EPHA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011540969.1 | XP_011539271.1 | 8, 9 | 1736 | ||
XM_011540970.1 | XP_011539272.1 | 6, 7 | 1470 | |||
XM_011540972.1 | XP_011539274.1 | 7, 8 | 1637 | |||
XM_011540973.1 | XP_011539275.1 | 5, 6 | 1372 | |||
XM_011540975.2 | XP_011539277.1 | 3, 4 | 548 | |||
GenBank mRNA | AB040892.1 | - | 4, 5 | 994 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
