siRNA Details
Gene Symbol: MMP28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024302.4 | NP_077278.1 | 8 | 2204 | ||
NM_032950.3 | NP_116568.1 | 9 | 2267 | |||
XM_011525225.1 | XP_011523527.1 | 7 | 1979 | |||
XM_011525226.2 | XP_011523528.1 | 9 | 2316 | |||
XM_011525227.1 | XP_011523529.1 | 9 | 1945 | |||
XM_011525228.1 | XP_011523530.1 | 9 | 1970 | |||
XM_011525229.2 | XP_011523531.1 | 8 | 1845 | |||
XM_011525230.1 | XP_011523532.1 | 8 | 2070 | |||
XM_011525232.2 | XP_011523534.2 | 8 | 2126 | |||
XM_017025061.1 | XP_016880550.1 | 9 | 1934 | |||
XM_017025062.1 | XP_016880551.1 | 8 | 2200 | |||
GenBank mRNA | AF330002.1 | - | 8 | 2149 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
