siRNA Details
Gene Symbol: EPHA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006943.1 | NP_001006944.1 | 3 | 490 | ||
XM_011540969.1 | XP_011539271.1 | 5 | 653 | |||
XM_011540970.1 | XP_011539272.1 | 3 | 387 | |||
XM_011540972.1 | XP_011539274.1 | 4 | 554 | |||
XM_011540973.1 | XP_011539275.1 | 2 | 289 | |||
GenBank mRNA | BC038796.1 | - | 3 | 420 | ||
BC072417.1 | - | 3 | 490 | |||
DR007053.1 | - | 3 | 500 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
