siRNA Details
Gene Symbol: KCNMA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161352.1 | NP_001154824.1 | 28 | 6018 | ||
NM_001161353.1 | NP_001154825.1 | 28 | 5967 | |||
NM_001271519.1 | NP_001258448.1 | 29 | 5934 | |||
NM_001322836.1 | NP_001309765.1 | 28 | 5853 | |||
NM_001322837.1 | NP_001309766.1 | 29 | 5937 | |||
NM_002247.3 | NP_002238.2 | 27 | 5844 | |||
XM_006717826.2 | XP_006717889.1 | 28 | 5846 | |||
XM_011539774.1 | XP_011538076.1 | 31 | 6145 | |||
XM_011539777.1 | XP_011538079.1 | 30 | 6100 | |||
XM_011539778.2 | XP_011538080.1 | 29 | 6020 | |||
XM_011539779.1 | XP_011538081.1 | 29 | 6052 | |||
XM_011539782.2 | XP_011538084.1 | 28 | 5917 | |||
XM_011539783.1 | XP_011538085.1 | 29 | 5932 | |||
XM_017016209.1 | XP_016871698.1 | 30 | 6220 | |||
XM_017016214.1 | XP_016871703.1 | 29 | 6046 | |||
XM_017016215.1 | XP_016871704.1 | 30 | 6013 | |||
XM_017016218.1 | XP_016871707.1 | 28 | 5383 | |||
XM_017016222.1 | XP_016871711.1 | 27 | 5833 | |||
GenBank mRNA | AK024857.1 | - | 1 | 1689 | ||
U11058.2 | - | 27 | 5558 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
