siRNA Details
Gene Symbol: CLCA3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024604.1 | 8, 9 | 1389 | |||
GenBank mRNA | AF043976.1 | - | 8, 9 | 1389 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CLCA3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024604.1 | 8, 9 | 1389 | |||
GenBank mRNA | AF043976.1 | - | 8, 9 | 1389 |