siRNA Details
Gene Symbol: PPP2R3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190447.1 | NP_001177376.1 | 9, 10 | 1105 | ||
NM_002718.4 | NP_002709.2 | 10, 11 | 3540 | |||
NM_181897.2 | NP_871626.1 | 9, 10 | 1395 | |||
XM_006713686.3 | XP_006713749.1 | 10, 11 | 3951 | |||
XM_011512956.2 | XP_011511258.1 | 10, 11 | 3715 | |||
XM_017006787.1 | XP_016862276.1 | 9, 10 | 3273 | |||
GenBank mRNA | AK293012.1 | - | 10, 11 | 3520 | ||
AK298059.1 | - | 9, 10 | 1034 | |||
AK299544.1 | - | 9, 10 | 1065 | |||
AK314243.1 | - | 10, 11 | 3468 | |||
AK316258.1 | - | 9, 10 | 1348 | |||
AL389975.1 | - | 7, 8 | 623 | |||
BC065531.1 | - | 10, 11 | 3487 | 6161260 | ||
L07590.1 | - | 10, 11 | 3427 | |||
L12146.1 | - | 9, 10 | 1320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
