siRNA Details
Gene Symbol: BSG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF548371.1 | - | 9 | 1392 | ||
BC009040.2 | - | 8 | 1028 | 3867352 | ||
D45131.1 | - | 8 | 1013 | |||
L20471.1 | - | 8 | 1029 | |||
M87879.1 | - | 8 | 1006 | |||
X64364.1 | - | 8 | 1041 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
