siRNA Details
Gene Symbol: TRPV5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019841.6 | NP_062815.3 | 15 | 2727 | ||
GenBank mRNA | AF304464.1 | - | 15 | 2812 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
