siRNA Details
Gene Symbol: CLK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020666.2 | NP_065717.1 | 4 | 526 | ||
GenBank mRNA | AF294429.1 | - | 3 | 418 | ||
AK304614.1 | - | 4 | 528 | |||
BC015942.1 | - | 4 | 526 | |||
BC136261.1 | - | 4 | 474 | 9020248 | ||
BC143547.1 | - | 4 | 474 | 9052058 | ||
BC151233.1 | - | 4 | 1740 | |||
CR749504.1 | - | 6 | 1405 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
