siRNA Details
Gene Symbol: CBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286789.1 | NP_001273718.1 | 3 | 1726 | ||
NM_001757.3 | NP_001748.1 | 3 | 1095 | |||
GenBank mRNA | AB124848.1 | - | 3 | 1095 | ||
AK311219.1 | - | 3 | 1626 | |||
BC002511.2 | - | 3 | 980 | 3050837 | ||
BC015640.2 | - | 3 | 994 | 4642240 | ||
J04056.1 | - | 3 | 996 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
