siRNA Details
Gene Symbol: KCNH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139318.4 | NP_647479.2 | 8 | 1548 | ||
NM_172375.2 | NP_758963.1 | 8 | 1548 | |||
GenBank mRNA | AF418206.1 | - | 8 | 1496 | ||
AF472412.1 | - | 8 | 1496 | |||
AF493798.1 | - | 8 | 1496 | |||
AK074484.1 | - | 8 | 1490 | |||
BC073979.1 | - | 8 | 1548 | 6199697 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
