siRNA Details
Gene Symbol: MTMR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017677.3 | NP_060147.2 | 12 | 1510 | ||
GenBank mRNA | AK000133.1 | - | 8 | 830 | ||
AK225231.1 | - | 8 | 830 | |||
BC012399.1 | - | 12 | 1488 | 3864128 |
