siRNA Details
Gene Symbol: CDK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261.3 | NP_001252.1 | 2 | 263 | ||
XM_017014184.1 | XP_016869673.1 | 2 | 129 | |||
GenBank mRNA | AB451289.1 | - | 2 | 140 | ||
AB451421.1 | - | 2 | 140 | |||
AK313828.1 | - | 2 | 192 | |||
BC001968.1 | - | 2 | 200 | 3460767 | ||
BT019903.1 | - | 2 | 140 | |||
DB120461.1 | - | 2 | 263 | |||
L25676.1 | - | 2 | 204 | |||
X80230.1 | - | 2 | 238 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
