siRNA Details
Gene Symbol: SENP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015670.5 | NP_056485.2 | 11 | 2482 | ||
GenBank mRNA | AF199459.1 | - | 12 | 2231 | ||
AL050283.1 | - | 11 | 1755 | |||
AL834294.1 | - | 12 | 2242 | |||
AY008763.1 | - | 12 | 2212 | |||
BC048306.1 | - | 10 | 1714 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
