siRNA Details
Gene Symbol: USP48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032236.5 | NP_115612.4 | 26, 27 | 3322 | ||
XM_005246009.2 | XP_005246066.1 | 25, 26 | 3170 | |||
XM_006710951.2 | XP_006711014.2 | 26, 27 | 3320 | |||
XM_006710955.3 | XP_006711018.1 | 25, 26 | 3104 | |||
XM_011542262.2 | XP_011540564.1 | 26, 27 | 3216 | |||
XM_011542263.2 | XP_011540565.1 | 26, 27 | 3216 | |||
XM_011542264.1 | XP_011540566.1 | 25, 26 | 2916 | |||
XM_011542265.2 | XP_011540567.1 | 27, 28 | 3882 | |||
XM_011542267.2 | XP_011540569.1 | 26, 27 | 3235 | |||
XM_011542268.2 | XP_011540570.1 | 18, 19 | 1935 | |||
XM_017002495.1 | XP_016857984.1 | 25, 26 | 3125 | |||
XM_017002497.1 | XP_016857986.1 | 18, 19 | 1980 | |||
GenBank mRNA | AF502942.1 | - | 26, 27 | 3224 | ||
AK002190.1 | - | 10, 11 | 953 | |||
AK026707.1 | - | 14, 15 | 1830 | |||
AK226150.1 | - | 25, 26 | 3120 | |||
AK315005.1 | - | 26, 27 | 3102 | |||
AL834375.1 | - | 11, 12 | 1015 | |||
BC013567.1 | - | 10, 11 | 1000 | |||
BC104896.1 | - | 25, 26 | 2962 | 8143899 | ||
BC143343.1 | - | 25, 26 | 2962 | 9051848 | ||
BC143349.1 | - | 26, 27 | 3118 | 9051854 | ||
BC143354.1 | - | 26, 27 | 3154 | 9051859 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Matching Taqman Gene Expression Assay:
Gene Ontology Categories:
