siRNA Details
Gene Symbol: MMP26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021801.4 | NP_068573.2 | 2 | 323 | ||
XM_011520219.2 | XP_011518521.1 | 3 | 503 | |||
GenBank mRNA | AF230354.1 | - | 2 | 307 | ||
AF248646.1 | - | 2 | 311 | |||
AF291664.1 | - | 2 | 320 | |||
AJ251531.1 | - | 2 | 317 | |||
BC101541.1 | - | 2 | 323 | 8069047 | ||
BC101543.1 | - | 2 | 323 | 8069049 |
