siRNA Details
Gene Symbol: RPS6KA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK295674.1 | - | 2 | 180 | ||
AK310428.1 | - | 2 | 99 | |||
BC002363.2 | - | 2, 4 | 332 | 2961482 | ||
BF205134.1 | - | 332 | ||||
DA320139.1 | - | 3 | 519 | |||
DC402728.1 | - | 2 | 112 | |||
KJ535087.1 | - | 3 | 137 | |||
L07598.1 | - | 2 | 117 | |||
X85106.1 | - | 2, 4 | 288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
