siRNA Details
Gene Symbol: RPS6KA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC002363.2 | - | 1, 4 | 325 | 2961482 | |
BF205134.1 | - | 325 | ||||
L07598.1 | - | 1, 2 | 110 | |||
X85106.1 | - | 1, 4 | 281 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Matching Taqman Gene Expression Assay:
