siRNA Details
Gene Symbol: CREM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011519316.2 | XP_011517618.1 | 4 | 402 | ||
GenBank mRNA | S68134.1 | - | 2 | 215 | ||
S68271.1 | - | 2 | 215 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
