siRNA Details
Gene Symbol: KLK13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015596.1 | NP_056411.1 | 4 | 607 | ||
GenBank mRNA | AB108823.1 | - | 2 | 108 | ||
AB108824.1 | - | 4 | 345 | |||
AB451465.1 | - | 4 | 564 | |||
AY923171.1 | - | 4 | 396 | |||
AY923172.1 | - | 5 | 963 | |||
AY923173.1 | - | 5 | 672 | |||
AY923174.1 | - | 4 | 879 | |||
AY923175.1 | - | 4 | 588 | |||
BC069334.1 | - | 4 | 608 | 7262205 | ||
BC069543.1 | - | 4 | 607 | 7262217 |
