siRNA Details
Gene Symbol: DIABLO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278342.1 | NP_001265271.1 | 2 | 658 | ||
NM_019887.5 | NP_063940.1 | 3 | 922 | |||
GenBank mRNA | AF262240.1 | - | 2 | 123 | ||
AK024768.1 | - | 2 | 131 | |||
AK222664.1 | - | 2 | 120 | |||
AK315629.1 | - | 2 | 142 | |||
AY313210.1 | - | 2 | 104 | |||
BC011909.2 | - | 2 | 127 | 4137792 | ||
BC046209.1 | - | 3 | 844 | |||
BF724951.1 | - | 2 | 125 | |||
KU178725.1 | - | 2 | 104 | |||
KU178726.1 | - | 2 | 104 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
