siRNA Details
Gene Symbol: PPP2R3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013239.4 | NP_037371.2 | 3 | 866 | ||
XM_011545630.2 | XP_011543932.1 | 3 | 866 | |||
GenBank mRNA | AF135016.1 | - | 2 | 230 | ||
AK293506.1 | - | 4 | 944 | |||
BC063429.1 | - | 3 | 621 | 5239974 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
