siRNA Details
Gene Symbol: PPP1R2P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002191.2 | 1 | 254 | |||
GenBank mRNA | AB044137.1 | - | 1 | 254 | ||
BC056673.1 | - | 1 | 184 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
