siRNA Details
Gene Symbol: ECI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001919.3 | NP_001910.2 | 5 | 555 | ||
GenBank mRNA | AK291127.1 | - | 5 | 555 | ||
BC000762.2 | - | 5 | 520 | 2821822 | ||
BC009631.1 | - | 6 | 497 | 3877066 | ||
BC019316.1 | - | 5 | 520 | 2821822 | ||
L24774.1 | - | 4 | 412 | |||
Z25820.1 | - | 5 | 528 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
