siRNA Details
Gene Symbol: DCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129889.2 | NP_001123361.1 | 1, 2 | 730 | ||
NM_001922.4 | NP_001913.2 | 1, 2 | 730 | |||
XM_017020401.1 | XP_016875890.1 | 1, 2 | 733 | |||
GenBank mRNA | AJ000503.1 | - | 1, 2 | 702 | ||
BC028311.1 | - | 1, 2 | 577 | 4705935 | ||
D17547.1 | - | 1, 2 | 717 | |||
DQ902581.1 | - | 1, 2 | 595 | |||
L18967.1 | - | 1, 2 | 687 | |||
S69231.1 | - | 1, 2 | 652 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
