siRNA Details
Gene Symbol: FANCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000135.2 | NP_000126.2 | 43 | 5359 | ||
NM_001286167.1 | NP_001273096.1 | 43 | 5363 | |||
GenBank mRNA | X99226.1 | - | 43 | 5347 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
