siRNA Details
Gene Symbol: CD44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000610.3 | NP_000601.3 | 11, 12 | 1851 | ||
NM_001001389.1 | NP_001001389.1 | 10, 11 | 1722 | |||
XM_005253231.2 | XP_005253288.1 | 11, 12 | 1581 | |||
XM_005253232.2 | XP_005253289.1 | 11, 12 | 1578 | |||
XM_005253233.2 | XP_005253290.1 | 10, 11 | 1461 | |||
XM_005253234.2 | XP_005253291.1 | 10, 11 | 1458 | |||
XM_005253235.2 | XP_005253292.1 | 10, 11 | 1452 | |||
XM_005253238.2 | XP_005253295.1 | 7, 8 | 1092 | |||
XM_006718388.1 | XP_006718451.1 | 11, 12 | 1575 | |||
XM_011520482.1 | XP_011518784.1 | 11, 12 | 1581 | |||
XM_011520483.1 | XP_011518785.1 | 9, 10 | 1326 | |||
XM_011520484.1 | XP_011518786.1 | 9, 10 | 1323 | |||
XM_011520485.1 | XP_011518787.1 | 9, 10 | 1320 | |||
XM_011520486.1 | XP_011518788.1 | 6, 7 | 963 | |||
GenBank mRNA | AJ251595.1 | - | 11, 12 | 1595 | ||
BC004372.1 | - | 10, 11 | 1419 | 3638681 | ||
BG946113.1 | - | 1, 2 | 97 | |||
M83328.1 | - | 2, 3 | 154 | |||
X62739.1 | - | 5, 6 | 621 | |||
X66733.1 | - | 10, 11 | 1417 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
