siRNA Details
Gene Symbol: GSR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000637.3 | NP_000628.2 | 10 | 1204 | ||
NM_001195102.1 | NP_001182031.1 | 9 | 1117 | |||
NM_001195103.1 | NP_001182032.1 | 9 | 1045 | |||
NM_001195104.1 | NP_001182033.1 | 8 | 958 | |||
GenBank mRNA | AB519179.1 | - | 9 | 983 | ||
AB519180.1 | - | 9 | 911 | |||
AB519181.1 | - | 8 | 824 | |||
AF228704.1 | - | 10 | 1070 | |||
BC035691.1 | - | 10 | 939 | |||
BC069244.1 | - | 10 | 1161 | 6422426 | ||
FJ224321.1 | - | 10 | 1161 | |||
GQ472231.1 | - | 10 | 1161 | |||
X15722.1 | - | 10 | 1045 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
