siRNA Details
Gene Symbol: BLVRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000712.3 | NP_000703.2 | 6 | 460 | ||
NM_001253823.1 | NP_001240752.1 | 7 | 554 | |||
XM_011515474.2 | XP_011513776.1 | 6 | 462 | |||
XM_017012520.1 | XP_016868009.1 | 6 | 469 | |||
GenBank mRNA | AK291862.1 | - | 7 | 541 | ||
BC005902.1 | - | 6 | 453 | 4282476 | ||
BC008456.1 | - | 6 | 453 | 4249842 | ||
U34877.1 | - | 6 | 433 | |||
X93086.1 | - | 6 | 437 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
