siRNA Details
Gene Symbol: CKS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001827.2 | NP_001818.1 | 3 | 402 | ||
GenBank mRNA | BC006458.1 | - | 3 | 368 | 3538689 | |
BM838320.1 | - | 3 | 402 | |||
X54942.1 | - | 3 | 399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
