siRNA Details
Gene Symbol: DBH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000787.3 | NP_000778.3 | 3 | 662 | ||
GenBank mRNA | BC017174.2 | - | 3 | 648 | 3163986 | |
CD014125.1 | - | 3 | 547 | |||
X13255.1 | - | 3 | 643 | |||
X13256.1 | - | 3 | 643 | |||
Y00096.1 | - | 3 | 650 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
