siRNA Details
Gene Symbol: HMGCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000859.2 | NP_000850.1 | 12 | 1708 | ||
NM_001130996.1 | NP_001124468.1 | 12 | 1708 | |||
XM_011543357.1 | XP_011541659.1 | 12 | 1859 | |||
XM_011543358.1 | XP_011541660.1 | 12 | 1746 | |||
XM_011543359.1 | XP_011541661.1 | 12 | 1859 | |||
GenBank mRNA | AK292892.1 | - | 12 | 1639 | ||
AK296499.1 | - | 12 | 1863 | |||
AK312437.1 | - | 11 | 1552 | |||
AY429541.1 | - | 1 | 94 | |||
BC033692.1 | - | 12 | 1644 | 5212903 | ||
M11058.1 | - | 12 | 1602 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
