siRNA Details
Gene Symbol: IFNAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289125.1 | NP_001276054.1 | 9 | 1839 | ||
NM_001289126.1 | NP_001276055.1 | 8 | 1662 | |||
NM_001289128.1 | NP_001276057.1 | 8 | 1708 | |||
NM_207585.2 | NP_997468.1 | 9 | 1793 | |||
GenBank mRNA | AK293059.1 | - | 8 | 1476 | ||
L41942.1 | - | 9 | 1760 | |||
L41943.1 | - | 7 | 1342 | |||
L41944.1 | - | 9 | 1992 | |||
U29584.1 | - | 9 | 1500 | |||
X89772.1 | - | 8 | 1434 | |||
X89814.1 | - | 8 | 1467 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
